siRNA Details
Gene Symbol: Zadh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146090.5 | NP_666202.2 | 1, 2 | 218 | ||
GenBank mRNA | AK047559.1 | - | 1, 2 | 223 | ||
AK081988.1 | - | 1, 2 | 219 | |||
AK083082.1 | - | 1, 2 | 262 | |||
BC116427.1 | - | 1, 2 | 219 | 40104327 | ||
BC116428.1 | - | 1, 2 | 219 | 40104329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
