siRNA Details
Gene Symbol: Pla2g16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139269.2 | NP_644675.2 | 4 | 378 | ||
GenBank mRNA | AB298806.1 | - | 2 | 223 | ||
AK039703.1 | - | 4 | 591 | |||
AK042007.1 | - | 4 | 331 | |||
AK052657.1 | - | 4 | 357 | |||
AK163505.1 | - | 4 | 379 | |||
AX775891.1 | - | 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
