siRNA Details
Gene Symbol: BC021614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144869.2 | NP_659118.1 | 4 | 291 | ||
GenBank mRNA | BC021614.1 | - | 4 | 291 | 5102505 | |
BC034269.1 | - | 5 | 304 | 4196879 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC021614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144869.2 | NP_659118.1 | 4 | 291 | ||
GenBank mRNA | BC021614.1 | - | 4 | 291 | 5102505 | |
BC034269.1 | - | 5 | 304 | 4196879 |