siRNA Details
Gene Symbol: Rorb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043354.2 | NP_001036819.1 | 4 | 1156 | ||
NM_001289921.1 | NP_001276850.1 | 5 | 675 | |||
NM_146095.4 | NP_666207.3 | 4 | 622 | |||
GenBank mRNA | AB258405.1 | - | 4 | 783 | ||
AK044421.1 | - | 4 | 617 | |||
AK134790.1 | - | 4 | 923 | |||
BC024768.1 | - | 5 | 658 | |||
BC024842.1 | - | 5 | 675 | 5358124 | ||
BC058269.1 | - | 3 | 529 | |||
DQ779923.1 | - | 3 | 526 | |||
DQ779924.1 | - | 4 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
