siRNA Details
Gene Symbol: Ermp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081213.1 | NP_001074682.1 | 2 | 467 | ||
GenBank mRNA | AK037287.1 | - | 2 | 426 | ||
AK041066.1 | - | 2 | 438 | |||
AK050284.1 | - | 2 | 463 | |||
AK129447.1 | - | 2 | 395 | |||
AK137206.1 | - | 2 | 423 | |||
BC086627.1 | - | 2 | 408 | |||
BC151181.1 | - | 2 | 426 | 9088070 | ||
BC151182.1 | - | 2 | 426 | 9088071 | ||
BC157914.1 | - | 2 | 426 | 9088152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
