siRNA Details
Gene Symbol: Cox15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144874.4 | NP_659123.2 | 7 | 1065 | ||
GenBank mRNA | AK048099.1 | - | 7 | 1047 | ||
AK051792.1 | - | 7 | 964 | |||
AK141631.1 | - | 7 | 997 | |||
BC011509.1 | - | 7 | 1025 | 3981950 | ||
BC021498.1 | - | 7 | 985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
