siRNA Details
Gene Symbol: Wbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177812.1 | NP_001171283.1 | 5 | 522 | ||
NM_001177813.1 | NP_001171284.1 | 4 | 465 | |||
NM_146099.3 | NP_666211.3 | 4 | 474 | |||
GenBank mRNA | AK052400.1 | - | 4 | 455 | ||
AK078491.1 | - | 4 | 466 | |||
AK087487.1 | - | 4 | 467 | |||
AK142695.1 | - | 4 | 466 | |||
AK154686.1 | - | 5 | 499 | |||
AK169294.1 | - | 4 | 556 | |||
BC026369.1 | - | 4 | 460 | 4160029 | ||
BC058949.1 | - | 4 | 429 | 5721290 |
