siRNA Details
Gene Symbol: Ablim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290813.1 | NP_001277742.1 | 1 | 184 | ||
NM_178688.3 | NP_848803.3 | 1 | 184 | |||
GenBank mRNA | AF404774.1 | - | 1 | 184 | ||
BC141083.1 | - | 1 | 178 | 9055695 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
