siRNA Details
Gene Symbol: Ablim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103177.2 | NP_001096647.1 | 14 | 1650 | ||
NM_001103178.2 | NP_001096648.1 | 12 | 1192 | |||
NM_001290813.1 | NP_001277742.1 | 14 | 1636 | |||
NM_001290815.1 | NP_001277744.1 | 14 | 1650 | |||
NM_001290816.1 | NP_001277745.1 | 13 | 1545 | |||
NM_178688.3 | NP_848803.3 | 17 | 1888 | |||
GenBank mRNA | AF404774.1 | - | 17 | 1888 | ||
AF404775.1 | - | 14 | 1410 | |||
AF404776.1 | - | 12 | 1152 | |||
AK122196.1 | - | 14 | 1650 | |||
AK143907.1 | - | 13 | 1315 | |||
AK171860.1 | - | 1442 | ||||
BC032216.1 | - | 4 | 340 | |||
BC141083.1 | - | 14 | 1630 | 9055695 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
