siRNA Details
Gene Symbol: Prlhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201615.2 | NP_963909.2 | 1 | 609 | ||
GenBank mRNA | BC125649.1 | - | 1 | 541 | 40130164 | |
BC125651.1 | - | 1 | 541 | 40130166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
