siRNA Details
Gene Symbol: Npbwr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010342.1 | NP_034472.1 | 1 | 978 | ||
GenBank mRNA | AK138357.1 | - | 1 | 983 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
