siRNA Details
Gene Symbol: Epb4.1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113416.1 | NP_001106887.1 | 12 | 1250 | ||
NM_145506.4 | NP_663481.2 | 12 | 1250 | |||
GenBank mRNA | AK034676.1 | - | 12 | 1172 | ||
AK037024.1 | - | 12 | 1206 | |||
AK040354.1 | - | 12 | 1218 | |||
AK044959.1 | - | 12 | 1188 | |||
AK044996.1 | - | 12 | 1196 | |||
AK047523.1 | - | 12 | 1177 | |||
AK079301.1 | - | 12 | 1201 | |||
AK136125.1 | - | 12 | 1205 | |||
AK161141.1 | - | 12 | 1201 | |||
AK167449.1 | - | 12 | 1250 | |||
AK173198.1 | - | 12 | 1184 | |||
BC003937.1 | - | 12 | 1235 | 3486543 | ||
BC011476.1 | - | 12 | 1184 | 4014009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
