siRNA Details
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178690.4 | NP_848805.2 | 13 | 1255 | ||
GenBank mRNA | AK076244.1 | - | 13 | 1219 | ||
AK076399.1 | - | 13 | 1238 | |||
AK080432.1 | - | 13 | 1237 | |||
AK134604.1 | - | 13 | 1237 | |||
AK164523.1 | - | 13 | 1236 | |||
AK170768.1 | - | 13 | 1228 | |||
AK172881.1 | - | 13 | 1242 | |||
BC046297.1 | - | 13 | 1221 | 6486170 |
