siRNA Details
Gene Symbol: Dars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177445.5 | NP_803228.2 | 11, 12 | 1303 | ||
GenBank mRNA | AK077533.1 | - | 11, 12 | 1226 | ||
AK144470.1 | - | 11, 12 | 1249 | |||
AK146350.1 | - | 11, 12 | 1247 | |||
AK151027.1 | - | 11, 12 | 1247 | |||
AK151897.1 | - | 11, 12 | 1214 | |||
AK152304.1 | - | 11, 12 | 1214 | |||
AK152562.1 | - | 11, 12 | 1214 | |||
AK153256.1 | - | 11, 12 | 1214 | |||
AK153299.1 | - | 11, 12 | 1214 | |||
AK166009.1 | - | 11, 12 | 1214 | |||
AK169247.1 | - | 11, 12 | 1214 | |||
AK169716.1 | - | 11, 12 | 1191 | |||
BC008638.1 | - | 11, 12 | 1201 | 3586278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
