siRNA Details
Gene Symbol: 5430435G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145509.2 | NP_663484.1 | 4 | 533 | ||
GenBank mRNA | AB232641.1 | - | 3 | 377 | ||
AK030688.1 | - | 4 | 533 | |||
AK036056.1 | - | 4 | 522 | |||
AK079999.1 | - | 4 | 501 | |||
CJ137398.1 | - | 4 | 452 | |||
JX431303.1 | - | 3 | 377 |
