siRNA Details
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153774.1 | NP_722469.1 | 4 | 415 | ||
GenBank mRNA | AF273672.1 | - | 4 | 360 | ||
AF273673.1 | - | 4 | 360 | |||
AJ309238.1 | - | 4 | 354 | |||
AK036687.1 | - | 4 | 365 | |||
AK173127.1 | - | 4 | 355 | |||
BC056469.1 | - | 4 | 382 | |||
BC079546.1 | - | 5 | 526 | |||
BC098508.1 | - | 4 | 365 | 30015513 | ||
CD347972.1 | - | 5 | 526 |
