siRNA Details
Gene Symbol: Rasal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177644.5 | NP_808312.3 | 7, 8 | 1589 | ||
GenBank mRNA | AK039584.1 | - | 7, 8 | 1589 | ||
AK086812.1 | - | 7, 8 | 1215 | |||
BC120902.2 | - | 7, 8 | 932 | 40086771 |
