siRNA Details
Gene Symbol: Dars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172644.3 | NP_766232.1 | 14 | 2063 | ||
GenBank mRNA | AK030952.1 | - | 14 | 2052 | ||
AK033634.1 | - | 13 | 2886 | |||
AK037337.1 | - | 14 | 2082 | |||
AK048454.1 | - | 14 | 2040 | |||
AK083300.1 | - | 3 | 472 | |||
AK138809.1 | - | 14 | 2064 | |||
AK154133.1 | - | 14 | 2091 | |||
AK154348.1 | - | 14 | 2092 | |||
AK164885.1 | - | 14 | 2034 | |||
BC046429.1 | - | 7 | 828 | |||
BC057943.1 | - | 13 | 2024 | |||
BC076606.1 | - | 14 | 2055 | 30625515 | ||
BC089191.1 | - | 14 | 2037 | 30473852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
