siRNA Details
Gene Symbol: Aph1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146104.3 | NP_666216.1 | 4 | 715 | ||
NM_146134.2 | NP_666246.1 | 4 | 715 | |||
GenBank mRNA | AK050828.1 | - | 2 | 938 | ||
AK078343.1 | - | 2 | 180 | |||
AK148400.1 | - | 4 | 668 | |||
AX775953.1 | - | 4 | 579 | |||
BC012406.1 | - | 4 | 640 | 3483214 | ||
BC024111.1 | - | 4 | 581 | 5098973 | ||
BC057865.1 | - | 4 | 658 | 5320175 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014255, XM_006232977, BC087081.
Gene Ontology Categories:
