siRNA Details
Gene Symbol: Prrc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081290.1 | NP_001074759.1 | 5 | 699 | ||
GenBank mRNA | AK007762.1 | - | 5 | 660 | ||
AK013732.1 | - | 5 | 525 | |||
AK030766.1 | - | 5 | 639 | |||
AK077748.1 | - | 5 | 525 | |||
AK082487.1 | - | 5 | 669 | |||
AK122441.1 | - | 5 | 612 | |||
AK135245.1 | - | 5 | 641 | |||
AK166509.1 | - | 5 | 688 | |||
AK167018.1 | - | 5 | 654 | |||
BC021412.1 | - | 5 | 627 | |||
BC053406.1 | - | 5 | 586 | |||
BC064009.1 | - | 5 | 608 | |||
BC080672.1 | - | 5 | 618 |

More Information
Other Species Targeted

Gene Ontology Categories:
