siRNA Details
Gene Symbol: Prrc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081290.1 | NP_001074759.1 | 2 | 346 | ||
GenBank mRNA | AK007762.1 | - | 2 | 307 | ||
AK013732.1 | - | 2 | 193 | |||
AK030766.1 | - | 2 | 286 | |||
AK077748.1 | - | 2 | 194 | |||
AK082487.1 | - | 2 | 316 | |||
AK122441.1 | - | 2 | 259 | |||
AK135245.1 | - | 2 | 288 | |||
AK166509.1 | - | 2 | 341 | |||
AK167018.1 | - | 2 | 307 | |||
BC006723.1 | - | 2 | 290 | |||
BC021412.1 | - | 2 | 274 | |||
BC037745.1 | - | 2 | 271 | |||
BC053406.1 | - | 2 | 239 | |||
BC064009.1 | - | 2 | 255 | |||
BC080672.1 | - | 2 | 271 |
