siRNA Details
Gene Symbol: AI607873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204910.1 | NP_001191839.1 | 7 | 3069 | ||
GenBank mRNA | BC150711.1 | - | 3084 | 9087622 | ||
BE335648.1 | - | 2 | 269 | |||
BX527645.1 | - | 1 | 45 |
