siRNA Details
Gene Symbol: C130074G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178692.3 | NP_848807.1 | 1 | 327 | ||
GenBank mRNA | AK052654.1 | - | 1 | 291 | ||
AK081755.1 | - | 1 | 306 | |||
AK084264.1 | - | 1 | 300 | |||
AK087640.1 | - | 1 | 318 | |||
BC116951.1 | - | 1 | 327 | 40126270 | ||
BC116953.1 | - | 1 | 327 | 40126272 | ||
BY710149.1 | - | 1 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
