siRNA Details
Gene Symbol: Slc30a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033286.2 | NP_001028458.1 | 4 | 1183 | ||
GenBank mRNA | AK053537.1 | - | 3 | 1858 | ||
AK136932.1 | - | 4 | 991 | |||
BC054548.1 | - | 3 | 1813 | |||
BC080293.1 | - | 2 | 259 | |||
BC108419.1 | - | 4 | 571 |
