siRNA Details
Gene Symbol: Plekhb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145516.2 | NP_663491.1 | 3 | 214 | ||
NM_175421.1 | NP_780630.1 | 3 | 214 | |||
GenBank mRNA | AF189817.1 | - | 3 | 174 | ||
AK039771.1 | - | 3 | 182 | |||
AK082118.1 | - | 3 | 167 | |||
AK148653.1 | - | 3 | 197 | |||
AK149868.1 | - | 3 | 174 | |||
AK150074.1 | - | 3 | 174 | |||
AK166091.1 | - | 3 | 182 | |||
AK179998.1 | - | 3 | 127 | |||
AK187330.1 | - | 127 | ||||
BC057994.1 | - | 3 | 182 | 5344583 | ||
CK625615.1 | - | 3 | 214 |
