siRNA Details
Gene Symbol: Hibch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146108.1 | NP_666220.1 | 12 | 1061 | ||
GenBank mRNA | AK076038.1 | - | 12 | 1060 | ||
AK082965.1 | - | 12 | 1061 | |||
BC026437.1 | - | 12 | 1061 | 4238681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
