siRNA Details
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153088.2 | NP_694728.1 | 3 | 550 | ||
GenBank mRNA | AK089478.1 | - | 3 | 458 | ||
AK167390.1 | - | 3 | 308 | |||
AY028804.1 | - | 3 | 575 | |||
BC049184.1 | - | 3 | 410 | 6439402 | ||
BC058085.1 | - | 2 | 737 | |||
BC065158.1 | - | 3 | 285 | 5717102 | ||
BC079638.1 | - | 3 | 285 | 5717102 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001128079, XM_006245236, BC161976.
Gene Ontology Categories:
