siRNA Details
Gene Symbol: Gpr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033290.2 | NP_001028462.2 | 2 | 1121 | ||
GenBank mRNA | AK146484.1 | - | 2 | 1077 | ||
BC115578.1 | - | 2 | 961 | 40102229 | ||
BC115579.1 | - | 2 | 961 | 40102231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
