siRNA Details
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 9 | 893 | ||
GenBank mRNA | AK050860.1 | - | 9 | 853 | ||
AK086699.1 | - | 9 | 887 | |||
AK171713.1 | - | 9 | 919 | |||
BC009153.1 | - | 9 | 882 | 2655209 | ||
BI739824.1 | - | 7 | 518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
