siRNA Details
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 11 | 1197 | ||
GenBank mRNA | AK050860.1 | - | 11 | 1157 | ||
AK086699.1 | - | 11 | 1191 | |||
AK171713.1 | - | 11 | 1223 | |||
BC009153.1 | - | 11 | 1186 | 2655209 |
