siRNA Details
Gene Symbol: Farp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145519.2 | NP_663494.2 | 8, 9 | 888 | ||
GenBank mRNA | AK050860.1 | - | 8, 9 | 848 | ||
AK086699.1 | - | 8, 9 | 882 | |||
AK171713.1 | - | 8, 9 | 914 | |||
BC009153.1 | - | 8, 9 | 877 | 2655209 | ||
BI739824.1 | - | 6, 7 | 513 |
