siRNA Details
Gene Symbol: Zcchc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122675.1 | NP_001116147.1 | 13 | 2956 | ||
NM_001122676.1 | NP_001116148.1 | 13 | 2956 | |||
GenBank mRNA | AK037100.1 | - | 7 | 1129 | ||
AK173248.1 | - | 13 | 2673 | |||
BC035528.1 | - | 10 | 1461 | |||
BC082306.1 | - | 9 | 1379 |
