siRNA Details
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110214.1 | NP_001103684.1 | 4 | 368 | ||
NM_146114.3 | NP_666226.2 | 4 | 368 | |||
NM_175683.4 | NP_783614.1 | 4 | 368 | |||
GenBank mRNA | AF387731.1 | - | 4 | 350 | ||
AK037126.1 | - | 4 | 335 | |||
AK038788.1 | - | 4 | 298 | |||
AK052369.1 | - | 4 | 324 | |||
AK052490.1 | - | 4 | 307 | |||
AK088810.1 | - | 3 | 251 | |||
AK166324.1 | - | 4 | 311 | |||
BC108935.2 | - | 4 | 321 | 40039591 | ||
BY729142.1 | - | 4 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
