siRNA Details
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110214.1 | NP_001103684.1 | 4 | 361 | ||
NM_146114.3 | NP_666226.2 | 4 | 361 | |||
NM_175683.4 | NP_783614.1 | 4 | 361 | |||
GenBank mRNA | AF387731.1 | - | 4 | 343 | ||
AK037126.1 | - | 4 | 328 | |||
AK038788.1 | - | 4 | 291 | |||
AK052369.1 | - | 4 | 317 | |||
AK052490.1 | - | 4 | 300 | |||
AK088810.1 | - | 3 | 244 | |||
AK166324.1 | - | 4 | 304 | |||
BC108935.2 | - | 4 | 314 | 40039591 | ||
BY729142.1 | - | 4 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
