siRNA Details
Gene Symbol: Tmem203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177344.3 | NP_796318.2 | 1 | 502 | ||
GenBank mRNA | AK050191.1 | - | 1 | 487 | ||
AK051471.1 | - | 1 | 436 | |||
AX775749.1 | - | 1 | 490 | |||
BC022606.1 | - | 1 | 470 | 4165766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
