siRNA Details
Gene Symbol: BC029214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153557.1 | NP_705785.1 | 5 | 667 | ||
GenBank mRNA | AK046066.1 | - | 4 | 597 | ||
AK053101.1 | - | 3 | 279 | |||
BC029214.1 | - | 5 | 667 | 4161391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
