siRNA Details
Gene Symbol: Lcn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100455.2 | NP_001093925.1 | 4 | 416 | ||
GenBank mRNA | AY301267.1 | - | 4 | 416 | ||
BE864901.1 | - | 3 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
