siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 7 | 544 | ||
AK086119.1 | - | 6 | 516 | |||
AK129355.1 | - | 1 | 14 | |||
AK147625.1 | - | 5 | 2493 | |||
BB662543.1 | - | 6 | 511 | |||
BC141190.1 | - | 6 | 589 | 9055802 | ||
BC171963.1 | - | 6 | 589 | 9054249 | ||
FJ628361.1 | - | 7 | 587 |
