siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 30, 31 | 3515 | ||
AK039085.1 | - | 13, 14 | 2878 | |||
AK046816.1 | - | 18, 19 | 2611 | |||
AK048406.1 | - | 12, 13 | 1393 | |||
AK081253.1 | - | 2, 3 | 140 | |||
AK129355.1 | - | 24, 25 | 2985 | |||
AK147625.1 | - | 28, 29 | 5464 | |||
BC057073.1 | - | 6, 7 | 1101 | |||
BC141190.1 | - | 29, 30 | 3560 | 9055802 | ||
BC171963.1 | - | 29, 30 | 3560 | 9054249 | ||
FJ628361.1 | - | 30, 31 | 3558 |

More Information
Other Species Targeted

