siRNA Details
Gene Symbol: Gca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145523.3 | NP_663498.1 | 6 | 596 | ||
GenBank mRNA | AB088388.1 | - | 6 | 490 | ||
AF518325.1 | - | 6 | 580 | |||
AK033740.1 | - | 6 | 629 | |||
AK046347.1 | - | 6 | 638 | |||
AK049317.1 | - | 6 | 597 | |||
AK050540.1 | - | 6 | 638 | |||
AK078915.1 | - | 6 | 597 | |||
BC021450.1 | - | 6 | 590 | 5044040 |
