siRNA Details
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087927.1 | - | 4 | 458 | ||
BC004636.1 | - | 4 | 449 | 3585719 | ||
BC057960.1 | - | 4 | 395 | 5354784 | ||
DQ230973.1 | - | 3 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
