siRNA Details
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145524.3 | NP_663499.2 | 4 | 552 | ||
GenBank mRNA | AK042669.1 | - | 4 | 530 | ||
AK087927.1 | - | 4 | 482 | |||
BC004636.1 | - | 4 | 473 | 3585719 | ||
BC057960.1 | - | 4 | 419 | 5354784 | ||
DQ093357.1 | - | 4 | 529 | |||
DQ093358.1 | - | 4 | 529 | |||
DQ230973.1 | - | 3 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
