siRNA Details
Gene Symbol: Mettl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110512.1 | NP_001103982.1 | 4, 5 | 419 | ||
NM_145524.3 | NP_663499.2 | 5, 6 | 883 | |||
GenBank mRNA | BC004636.1 | - | 5, 6 | 804 | 3585719 | |
BC057960.1 | - | 5, 6 | 750 | 5354784 | ||
DQ093358.1 | - | 5, 6 | 860 | |||
DQ230973.1 | - | 4, 5 | 651 | |||
DQ230974.1 | - | 4, 5 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
