siRNA Details
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172666.3 | NP_766254.2 | 19, 20 | 1935 | ||
GenBank mRNA | AK031049.1 | - | 19, 20 | 1937 | ||
AK031154.1 | - | 19, 20 | 1776 | |||
AK079447.1 | - | 18, 19 | 1615 | |||
AK164922.1 | - | 6, 7 | 417 | |||
BC063086.1 | - | 19, 20 | 1920 | 6332574 |
