siRNA Details
Gene Symbol: Gm561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033297.2 | NP_001028469.1 | 2 | 210 | ||
GenBank mRNA | AK002610.1 | - | 2 | 211 | ||
AK010324.1 | - | 2 | 193 | |||
BC147042.1 | - | 2 | 205 | 9056050 | ||
BC147043.1 | - | 2 | 205 | 9056078 |
