siRNA Details
Gene Symbol: Nkx2-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023504.1 | NP_075993.1 | 2 | 727 | ||
GenBank mRNA | AF202039.1 | - | 2 | 524 | ||
AK005620.1 | - | 1 | 357 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nkx2-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023504.1 | NP_075993.1 | 2 | 727 | ||
GenBank mRNA | AF202039.1 | - | 2 | 524 | ||
AK005620.1 | - | 1 | 357 |