siRNA Details
Gene Symbol: Cstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177655.3 | NP_808323.1 | 2 | 321 | ||
GenBank mRNA | AK131797.1 | - | 2 | 322 | ||
AK132807.1 | - | 2 | 324 | |||
AY707915.1 | - | 2 | 321 | |||
BC048646.1 | - | 2 | 321 | 6772392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
