siRNA Details
Gene Symbol: Nrsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009948.1 | NP_001009948.1 | 2 | 95 | ||
GenBank mRNA | AK140190.1 | - | 2 | 114 | ||
BC088982.1 | - | 2 | 127 | 5700110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
