siRNA Details
Gene Symbol: 6820408C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289738.1 | NP_001276667.1 | 8 | 1065 | ||
NM_177656.4 | NP_808324.1 | 8 | 1062 | |||
GenBank mRNA | AK078482.1 | - | 8 | 1062 | ||
BC137826.1 | - | 7 | 942 | 8860845 | ||
BC144943.1 | - | 7 | 939 | |||
BC144944.1 | - | 7 | 942 | 9053492 | ||
BC144945.1 | - | 6 | 835 |
