siRNA Details
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199303.2 | NP_955007.2 | 6 | 468 | ||
GenBank mRNA | BC048083.1 | - | 6 | 468 | 6312228 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bpifb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199303.2 | NP_955007.2 | 6 | 468 | ||
GenBank mRNA | BC048083.1 | - | 6 | 468 | 6312228 |